Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Dimethyl-2,2'-bipyridyl 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

OR13729-25G OR13729-1G OR13729-5G
76566-562EA 19.57 USD InStock
76566-562 76566-564 76566-566
4,4'-Dimethyl-2,2'-bipyridyl 99%
4,4'-Dimethyl-2,2'-bipyridyl


Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 169…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00006441
CAS Number: 1134-35-6
EINECS: 214-483-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty