Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(4-Methylpiperazino)-2-nitroaniline

Supplier: APOLLO SCIENTIFIC
Synonyms: 5-(4-Methylpiperazin-1-yl)-2-nitroaniline

OR13864-1G OR13864-5G
76566-602EA 46 USD InStock
76566-602 76566-600
5-(4-Methylpiperazino)-2-nitroaniline
5-(4-Methylpiperazino)-2-nitroaniline


Formula: C₁₁H₁₆N₄O₂
MW: 236.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00545964
CAS Number: 23491-48-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty