Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-amino-3-hydroxybenzoate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 4-amino-3-hydroxybenzoate

OR14134-5G OR14134-25G OR14134-1G
76566-738EA 13.33 USD InStock
76566-738 76566-740 76566-742
Methyl-4-amino-3-hydroxybenzoate
Methyl 4-amino-3-hydroxybenzoate


Formula: C₈H₉NO₃
MW: 167.17 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00017093
CAS Number: 63435-16-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty