Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR14137-1G OR14137-5G OR14137-500MG
76566-746EA 113 USD InStock
76566-746 76566-748 76566-750
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate


Formula: C₁₀H₈ClNO₃
MW: 225.63 g/mol
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD08689722
CAS Number: 924862-18-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty