Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazino-1H-1,3-benzimidazole

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Hydrazinyl-1H-benzo[d]imidazole

OR14212-1G OR14212-25G OR14212-5G
76566-848EA 19.48 USD InStock
76566-848 76566-850 76566-852
2-Hydrazino-1H-1,3-benzimidazole
2-Hydrazino-1H-1,3-benzimidazole


Formula: C₇H₈N₄
MW: 148.17 g/mol
Melting Pt: 224…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00513603
CAS Number: 15108-18-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty