Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-prolinol

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-1-Boc-2-pyrrolidinemethanol(S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanolN-Boc-L-ProlinolN-(tert-Butoxycarbonyl)-L-prolinol

OR14516-100G OR14516-25G OR14516-5G
76567-104EA 17 USD InStock
76567-104 76567-102 76567-106
N-Boc-L-prolinol
N-Boc-L-prolinol


Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Boiling Pt: 150 °C/4mmHg
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00066232
CAS Number: 69610-40-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty