Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-3-(1-naphthyl)-L-alanine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Fmoc-1-Nal-OH

OR14596-10G OR14596-5G
76560-758EA 108 USD InStock
76560-758 76561-078
Fmoc-3-(1-naphthyl)-L-alanine 98%
Fmoc-3-(1-naphthyl)-L-alanine


Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151914
CAS Number: 96402-49-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty