Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-formylpyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 6-Bromopyridine-2-carboxaldehyde6-Bromopyridine-2-carbaldehyde6-Bromo-2-pyridinecarboxaldehyde

OR1460-100G OR1460-25G
76567-136EA 176 USD InStock
76567-136 76567-134
2-Bromo-6-formylpyridine 95%
2-Bromo-6-formylpyridine


Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 79…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty