Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3-Pyridyl)-L-alanine 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-2-Amino-3-(pyridin-3-yl)propanoic acidH-β-(3-Pyridyl)-Ala-OHH-3-Pal-OH

OR14723-1G OR14723-5G
76567-176EA 469 USD InStock
76567-176 76567-174
3-(3-Pyridyl)-L-alanine 95%
3-(3-Pyridyl)-L-alanine


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 248…252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079676
CAS Number: 64090-98-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty