Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 2-amino-1,3-thiazole-4-carboxylate hydrobromide

OR14763-5G OR14763-1G
76567-220EA 14 USD InStock
76567-220 76567-218
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide


Formula: C₆H₉BrN₂O₂S
MW: 253.12 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06653384
CAS Number: 127942-30-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty