Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-alanine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Boc-Ala-OHN-Alpha-t-Boc-L-alanineBOC-L-Alanine(S)-2-((tert-Butoxycarbonyl)amino)propanoic acid

OR14778-250G OR14778-500G OR14778-100G
76567-236EA 18 USD InStock
76567-236 76567-240 76567-238
N-Boc-L-alanine
N-Boc-L-alanine


Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Melting Pt: 79…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00037225
CAS Number: 15761-38-3
EINECS: 239-847-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty