Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-4-bromobutyrate 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 4-bromobutyrate

OR14983-500G OR14983-100G OR14983-1KG
76567-340EA 177 USD InStock
76567-340 76567-338 76567-336
Ethyl-4-bromobutyrate 97%
Ethyl 4-bromobutyrate


Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 80…82 °C/10mmHg
Density: 1.363 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000259
CAS Number: 2969-81-5
EINECS: 221-005-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty