Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-[(Benzoylcarbamothioyl)amino]-m-xylene1-Benzoyl-3-(3,5-dimethylphenyl)thiourea

OR15025-5G
76567-388EA 27 USD InStock
76567-388
N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide
N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide


Formula: C₁₆H₁₆N₂OS
MW: 284.376 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD02631870
CAS Number: 117174-79-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty