Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-amino-3-nitrobenzoate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 4-amino-3-nitrobenzoate

OR15213-100G OR15213-25G OR15213-5G
76567-566EA 174 USD InStock
76567-566 76567-564 76567-562
Methyl-4-amino-3-nitrobenzoate
Methyl 4-amino-3-nitrobenzoate


Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00017562
CAS Number: 3987-92-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty