Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-4-(2-chloroethyl)piperazine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-(2-Chloroethyl)piperazine,N1-BOC protectedtert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

OR15245-10G OR15245-1G OR15245-5G
76567-588EA 13 USD InStock
76567-588 76567-592 76567-590
1-Boc-4-(2-chloroethyl)piperazine
1-Boc-4-(2-chloroethyl)piperazine


Formula: C₁₁H₂₁ClN₂O₂
MW: 248.751 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08443993
CAS Number: 208167-83-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty