Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Baicalein

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5,6,7-Trihydroxy-2-phenylchromen-4-one5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one5,6,7-Trihydroxyflavone

OR1525T-5G OR1525T-1G
76567-624EA 14 USD InStock
76567-624 76567-622
Baicalein
Baicalein
An inhibitor of 12-lipoxygenase, leukotriene biosynthesis & release of lysosomal enzymes. It also inhibits cellular calcium-ion uptake, mobilization & adjuvant-induced arthirritis.


Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 264…265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017459
CAS Number: 491-67-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty