Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2,6-dimethylpyridine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,6-Dimethylpyridin-4-amine

OR15746-1G OR15746-5G
76567-938EA 119 USD InStock
76567-938 76567-936
4-Amino-2,6-dimethylpyridine
4-Amino-2,6-dimethylpyridine


Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Melting Pt: 186…187 °C
Density: 1.039 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130078
CAS Number: 3512-80-9
EINECS: 225-515-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty