Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-3-iodophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR16668-250MG OR16668-1G
76568-262EA 32 USD InStock
76568-262 76568-264
4-Bromo-3-iodophenol 95%
4-Bromo-3-iodophenol


Formula: C₆H₄BrIO
MW: 298.904 g/mol
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00070745
CAS Number: 202865-84-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty