Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Hexahydrocyclopenta[c]pyrrol-4(1H)-one, N-BOC protected

OR16703-250MG OR16703-1G
76568-310EA 67 USD InStock
76568-310
4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester 98%
4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester


Formula: C₁₂H₁₉NO₃
MW: 225.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658358
CAS Number: 879686-42-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty