Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Oxa-1-azaspiro[4.4]nonane hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR16710-250MG OR16710-1G
76568-342EA 372 USD InStock
76568-342 76568-344
7-Oxa-1-azaspiro[4.4]nonane hydrochloride
7-Oxa-1-azaspiro[4.4]nonane hydrochloride


Formula: C₇H₁₃NO·ClH
MW: 163.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26960527
CAS Number: 1620569-18-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty