Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maleic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR17920-25G OR17920-10G OR17920-100G
76568-916EA 38 USD InStock
76568-916 76568-918 76568-920
Maleic acid
Maleic acid


Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 160 °C
Melting Pt: 132…140 °C
Density: 1.59 g/cm³ (25 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty