Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guaifenesin 99+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-(2-Methoxyphenoxy)-1,2-propanediol3-(2-Methoxyphenoxy)propane-1,2-diolGuaiacol glyceryl ether

OR18315-1KG
76569-188EA 91 USD InStock
76569-188
Guaifenesin 99+%
Guaifenesin


Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 215 °C/15mmHg
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00016873
CAS Number: 93-14-1
EINECS: 202-222-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty