Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,8-Diaminonaphthalene 90%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Naphthalene-1,8-diamine1,8-Naphthalenediamine1,8-naphthylenediamine

OR18317-100G OR18317-250G OR18317-1KG
76569-194EA 221 USD InStock
76569-194 76569-192 76569-190
1,8-Diaminonaphthalene 90%
1,8-Diaminonaphthalene


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 205 °C/12mmHg
Melting Pt: 62…64 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty