Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Trimethoxybenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: O,O,O-1,3,5-Trimethylresorcinol

OR18342-100G OR18342-25G
76569-274EA 29 USD InStock
76569-274 76569-272
1,3,5-Trimethoxybenzene 98%
1,3,5-Trimethoxybenzene


Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 225 °C
Melting Pt: 51…53 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008385
CAS Number: 621-23-8
EINECS: 210-673-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty