Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Hydroxyphenylazo)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4'-Hydroxyazobenzene-2-carboxylic acidHABA2-(4-Hydroxyphenylazo)benzoic acid4-Hydroxyazobenzene-2'-carboxylic acid

OR18374-25G OR18374-100G
76569-418EA 61 USD InStock
76569-418 76569-416
2-(4-Hydroxyphenylazo)benzoic acid
2-(4-Hydroxyphenylazo)benzoic acid


Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00002428
CAS Number: 1634-82-8
EINECS: 216-655-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty