Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoaniline 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR1857-25G OR1857-100G
76569-518EA 72 USD InStock
76569-518 76569-516
2-Iodoaniline 98+%
2-Iodoaniline


Formula: C₆H₆IN
MW: 219.02 g/mol
Melting Pt: 53…58 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty