Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Chlorodracyclic acidChlorodracylic acidp-Chlorobenzoic acid

OR1870-1KG OR1870-500G
76569-586EA 33 USD InStock
76569-586 76569-584
4-Chlorobenzoic acid
4-Chlorobenzoic acid


Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 275 °C
Melting Pt: 238…241 °C
Density: 1.54 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002531
CAS Number: 74-11-3
EINECS: 200-805-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty