Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-5-nitroanisole 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Chloro-3-methoxy-5-nitrobenzene3-Chloro-5-nitrophenyl methyl ether

OR18805-5G OR18805-25G OR18805-1G
76569-618EA 206 USD InStock
76569-618 76569-616 76569-614
3-Chloro-5-nitroanisole 95%
3-Chloro-5-nitroanisole


Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848441
CAS Number: 55910-07-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty