Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminonicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Amino-3-pyridinecarboxylic acid

OR1915-5G OR1915-100G OR1915-25G
76569-732EA 35 USD InStock
76569-732 76569-730 76569-734
5-Aminonicotinic acid
5-Aminonicotinic acid


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00129116
CAS Number: 24242-19-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty