Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl 4-(methoxymethyl)phenylacetate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-(Methoxymethyl)phenylacetic acid methyl ester

OR19481-1G OR19481-5G
76569-788EA 215 USD InStock
76569-788 76569-786
Methyl 4-(methoxymethyl)phenylacetate
Methyl 4-(methoxymethyl)phenylacetate


Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12674804
CAS Number: 115414-80-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty