Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3,5-dichlorobenzene

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3,5-Dichlorobromobenzene

OR19527-25G OR19527-250G OR19527-100G
76569-810EA 13.33 USD InStock
76569-810 76569-814 76569-812
1-Bromo-3,5-dichlorobenzene
1-Bromo-3,5-dichlorobenzene


Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 232 °C
Melting Pt: 73…77 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty