Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodostyrene

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Ethenyl-3-iodobenzene1-Iodo-3-vinylbenzene

OR19553-250MG OR19553-1G
76569-854EA 303 USD InStock
76569-854 76569-852
3-Iodostyrene
3-Iodostyrene


Formula: C₈H₇I
MW: 230.046 g/mol
Boiling Pt: 71…73 °C (3 mm Hg)
Density: 1.674 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD28103326
CAS Number: 4840-92-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty