Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitro-1,10-phenanthroline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR19583-5G OR19583-25G OR19583-1G
76569-872EA 128 USD InStock
76569-872 76569-870 76569-868
5-Nitro-1,10-phenanthroline 98%
5-Nitro-1,10-phenanthroline


Formula: C₁₂H₇N₃O₂
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS Number: 4199-88-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty