Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-chlorobenzene 98+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Bromochlorobenzene

OR1960-1KG OR1960-5KG OR1960-250G
76569-876EA 48 USD InStock
76569-876 76569-874 76569-878
1-Bromo-4-chlorobenzene 98+%
1-Bromo-4-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C
Melting Pt: 63…67 °C
Density: 1.651 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty