Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-N-ethylglycine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: N-Ethylglycine, N-BOC protected

OR20016-25G OR20016-5G OR20016-1G
76569-952EA 16 USD InStock
76569-952 76569-956 76569-954
N-Boc-N-ethylglycine
N-Boc-N-ethylglycine


Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Melting Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00672508
CAS Number: 149794-10-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty