Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dibromo-4-picoline

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,5-Dibromo-4-methylpyridine

OR2087-100G OR2087-25G OR2087-5G
76570-086EA 13.33 USD InStock
76570-086 76570-088 76570-090
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline


Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 36…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00234955
CAS Number: 3430-26-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty