Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-amino-2-methoxybenzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Amino-2-methoxybenzoic acid methyl esterMethyl 4-amino-2-methoxybenzoateMethyl 4-amino-2-methoxybenzenecarboxylate

OR21849-500G OR21849-100G OR21849-25G
76570-388EA 16 USD InStock
76570-388 76570-390 76570-392
Methyl-4-amino-2-methoxybenzoate 98%
Methyl 4-amino-2-methoxybenzoate


Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00017202
CAS Number: 27492-84-8
EINECS: 248-494-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty