Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,8-Bis(dimethylamino)naphthalene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine

OR21927-100G OR21927-10G OR21927-25G
76570-442EA 16 USD InStock
76570-442 76570-444 76570-446
1,8-Bis(dimethylamino)naphthalene 98%
1,8-Bis(dimethylamino)naphthalene


Formula: C₁₄H₁₈N₂
MW: 214.31 g/mol
Boiling Pt: 144…145 °C/7mmHg
Melting Pt: 49…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty