Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(1H-1,2,4-Triazol-1-yl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-[1,2,4]Triazol-1-yl-benzoic acid

OR23304-1G OR23304-250MG
76562-632EA 52 USD InStock
76562-632 76563-084
4-(1H-1,2,4-Triazol-1-yl)benzoic acid
4-(1H-1,2,4-Triazol-1-yl)benzoic acid


Formula: C₉H₇N₃O₂
MW: 189.1708 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682060
CAS Number: 162848-16-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty