Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

OR24192-5G OR24192-1G OR24192-25G
76571-278EA 94.76 USD InStock
76571-278 76571-276 76571-274
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate


Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00051652
CAS Number: 31037-02-2
EINECS: 31037-02-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty