Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylboronic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Phenyl boric acidDihydroxy(phenyl)boranePhenylmetaboric acidBenzeneboronic acid

OR2447-25G OR2447-100G
76571-386EA 48 USD InStock
76571-386 76571-384
Phenylboronic acid 99%
Phenylboronic acid


Formula: C₆H₇BO₂
MW: 121.93 g/mol
Melting Pt: 216…219 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty