Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-N,N-dimethylbenzamide

Supplier: APOLLO SCIENTIFIC
Warning

OR2638-10G OR2638-1G OR2638-5G
76562-080EA 276 USD InStock
76562-080 76559-992 76560-564
4-Amino-N,N-dimethylbenzamide
4-Amino-N,N-dimethylbenzamide


Formula: C₉H₁₂N₂O
MW: 164.21 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00523648
CAS Number: 6331-71-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty