Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR2857-100G OR2857-5G OR2857-25G
76572-456EA 41 USD InStock
76572-456 76572-458 76572-454
4-Amino-3-hydroxybenzoic acid
4-Amino-3-hydroxybenzoic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty