Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR28602-10G OR28602-25G OR28602-5G
76572-500EA 61 USD InStock
76572-500 76572-502 76572-498
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid


Formula: C₁₁H₇Cl₂NO₃
Melting Pt: 221…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00051658
CAS Number: 3919-76-4
EINECS: 223-487-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty