Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dimethoxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR28682-25G OR28682-100G
76572-580EA 19.57 USD InStock
76572-580 76572-582
2,4-Dimethoxybenzaldehyde
2,4-Dimethoxybenzaldehyde


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 165 °C/10mmHg
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003311
CAS Number: 613-45-6
EINECS: 210-342-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty