Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sesamol-(3,4-(methylenedioxy)phenol)

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Benzo[d][1,3]dioxol-5-olSesamol3,4-Methylenedioxyphenol

OR28697-100G OR28697-25G OR28697-5G
76572-602EA 53 USD InStock
76572-602 76572-598 76572-600
Sesamol-(3,4-(methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 113…116 °C/2mmHg
Melting Pt: 61…65 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005827
CAS Number: 533-31-3
EINECS: 208-561-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty