Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylpyrazine-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR28728-25G OR28728-100G OR28728-5G
76572-620EA 14 USD InStock
76572-620 76572-622 76572-618
5-Methylpyrazine-2-carboxylic acid
5-Methylpyrazine-2-carboxylic acid


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 165…171 °C
Storage Temperature: Ambient
EINECS: 413-260-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty