Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dihydroxy-3-cyclobutene-1,2-dione

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1,2-Dihydroxy-3,4-cyclobutenedioneSquaric acidQuadratic acid1,2-Dihydroxy-1-cyclobutene-3,4-dioneDiketocyclobutenediol

OR28821-100G OR28821-500G OR28821-25G
76572-686EA 473 USD InStock
76572-686 76572-682 76572-684
3,4-Dihydroxy-3-cyclobutene-1,2-dione
3,4-Dihydroxy-3-cyclobutene-1,2-dione


Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty