Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Chloropropiophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Chloropropiophenone

OR2891-500G OR2891-100G OR2891-25G
76572-756EA 13.33 USD InStock
76572-756 76572-758 76572-760
β-Chloropropiophenone 98%
β-Chloropropiophenone


Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168.62 g/mol
Boiling Pt: 113…115 °C/4mmHg
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty