Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Hydroxyphenyl)ethanol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: p-HPEANSC 59876Tyrosol4-Hydroxyphenethyl alcohol

OR2934-5G OR2934-25G OR2934-100G
76572-902EA 13.33 USD InStock
76572-902 76572-906 76572-904
2-(4-Hydroxyphenyl)ethanol 98%
2-(4-Hydroxyphenyl)ethanol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 139 °C/1mmHg
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty